Learn More
Waters Limited XSelect HSS T3 Sentry Guard Cartridge, 100, 3.5 m, 4.6 mm X 20 mm, 2/pk
Catalog No. 50466104
Encompass
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.
Catalog No. 50466104 Supplier Waters Limited Supplier No. 186004787
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.