Learn More
ACROBiosystems Inc Recombinant Protein;Human HSP90AA1 / HSP86 Protein, GST Tag;E.coli;1MG;HS1,HSP90AA1,HSP90A,HSP 86,HSP86,HSPC1,HSPCA,EL52,HSP89A,HSP90N,HSPCAL1,HSPCAL4,HSPN,Hsp89,Hsp90,LAP2
Catalog No. 501658522
Encompass
Recombinant Protein;Human HSP90AA1 / HSP86 Protein, GST Tag;1MG;Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It contains AA Glu 535 - Asp 732 (Accession # NP_005339).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 49.3 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 501658522 Supplier ACROBiosystems Inc Supplier No. HS1H513X1MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human HSP90AA1 / HSP86 Protein, GST Tag;1MG;Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It contains AA Glu 535 - Asp 732 (Accession # NP_005339).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 49.3 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE).