Learn More
Enzo Life Sciences [pSer326]HSF1 polyclonal antibody (50 g) DFS Item
Supplier:  Enzo ADISPA902D
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the N-terminus of human HSF1 phosphorylated at Ser 326. UniProt: Q00613. Source: Purified from rabbit serum. Species Reactivity: Human. Applications: WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50950114
Only null left
Explore available promotions