Learn More
Applied Biosystems™ HRM Software Module for QuantStudio™ 6 and 7 Systems
Catalog No. 4486395
Encompass
Change view
Click to view available options
Quantity:
10 licenses
Catalog No. Quantity
4486395 10 licenses
1 options
Catalog No. 4486395 Supplier Applied Biosystems™ Supplier No. 4486395
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This software module expands the functionality of the QuantStudio™ 6 Flex and QuantStudio™ 7 Flex real-time PCR system software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
High Resolution Melt (HRM) analysis is a new, post-PCR analysis method used for identifying genetic variation in nucleic acid sequences. Simple and fast, this method is based on PCR melt (dissociation) curve techniques and is enabled by the recent availability of improved double-stranded DNA (dsDNA)-binding dyes along with next-generation real-time PCR instrumentation and analysis software. HRM analysis can discriminate DNA sequences based on their composition, length, GC content, or strand complementarity.
Specifications
For Use With (Equipment) QuantStudio™ 6 Flex, QuantStudio™ 7
Product Line QuantStudio
Software Type HRM Analysis Software
Product Type Software Module
For Use With (Application) Real Time PCR (qPCR)
Quantity 10 licenses
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
What is the last software version for the ViiA7 Software?
ViiA7 Software v1.2.4 was the last version of the ViiA7 Software released. Currently, the ViiA7 Real-Time PCR System can be controlled using the QuantStudio Software. To obtain the most recent copy of the QuantStudio Software for performing experiments on the ViiA7 Real-Time PCR System, please visit the instrument software download page, https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/applied-biosystems-viia-7-real-time-pcr-system.html.