Learn More
Abcam Inc HPTS, Fluorescent membrane-impermeant pH indicator, 5G
Catalog No. 502705216
Encompass
MW 524.39 g/mol, Purity >98%. Fluorescent membrane-impermeant pH indicator. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705216 Supplier Abcam Inc Supplier No. AB1454135G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 524.39 g/mol, Purity >98%. Fluorescent membrane-impermeant pH indicator. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement