Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, Standard Size Replacement Chemical Exhaust Filter, Activated Carbon, 5/PK
Supplier:  Foxx Life Sciences 3300907OEM
Encompass
330-0907-OEM: EZwaste Chemical Exhaust Filter, 5pk. EZwaste Solvent Waste Filter Replacements come in packs of 1, 2, or 5. The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal of vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety. * Depending on application, carbon exhaust filter life cycle may vary
Catalog No. 501049967