missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dNTP Mix (20 mM)

Catalog No. AB0196
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0196 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0196 Supplier Thermo Scientific™ Supplier No. AB0196
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ dNTP Mix (20 mM) contains purified deoxyribonucleotide triphosphates (dNTPs) that meet high quality standards that result in excellent performance for a variety of molecular biology applications.

Thermo Scientific™ dNTP Mix (20 mM) contains purified deoxyribonucleotide triphosphates (dNTPs) that meet high quality standards that result in excellent performance for a variety of molecular biology applications. This pre-mixed solution contains 5 mM each of dATP, dCTP, dGTP, and dTTP at pH 7.5 for a total dNTP concentration of 20 mM. Such a mix makes it possible to reduce the total number of pipetting steps and the risk of reaction set-up errors.

Features

  • Purity confirmed by HPLC
  • Stable for 2 years at –20°C
  • Free of human and E. coli DNA

Applications
Thermo Scientific™ dNTP mix may be used in a wide variety of molecular applications including:

  • cDNA synthesis
  • PCR, RT-PCR, qPCR
  • Primer extension

Specifications

Concentration 20 mM
Content And Storage dNTP Mix (20 mM) contains premixed dATP, dCTP, dGTP and dTTP at a final concentration of 20 mM (1 mL)

Store at –20°C.
Form Liquid
Label or Dye Unlabeled
Quantity 1 mL
Shipping Condition Approved for shipment on Wet or Dry Ice
For Use With (Application) Standard PCR, Real Time PCR (qPCR)

For Research Use Only. Not for use in diagnostic procedures.