Learn More
Sterlitech Corporation Sterlitech HP4750 High Pressure Stirred Cell (S/P: HP4750)
Catalog No. 501952338
Encompass
The Sterlitech HP4750 stirred cell is a high-pressure stirred cell that is both chemically resistant and features a low hold-up volume (1 ml). This particular brand of stirred cell is able to perform a wide variety of membrane separations.
HP4750 Stirred Cell Product Benefits:
  • Max operating pressure is 1000 psi
  • Removable PTFE stir bar
  • 316 stainless steel
Catalog No. 501952338 Supplier Sterlitech Corporation Supplier No. HP4750
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Sterlitech HP4750 stirred cell is a high-pressure stirred cell that is both chemically resistant and features a low hold-up volume (1 ml). This particular brand of stirred cell is able to perform a wide variety of membrane separations.
HP4750 Stirred Cell Product Benefits:
  • Max operating pressure is 1000 psi
  • Removable PTFE stir bar
  • 316 stainless steel