missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Howard Leight™ QM24+™ Ear Muff

Catalog No. 19813222
Change view
Click to view available options
Noise Reduction Rating (NRR):
25 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Noise Reduction Rating (NRR)
19813222 25 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19813222 Supplier Honeywell™ Supplier No. QM24PLUS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The dielectric QM24+™ is an ultra-lightweight; three-position ear muff designed for extended wear at a valued price.

  • Crafted from tough ABS plastic
  • Designed for comfort during prolonged usage
  • Highly visible red cups are dielectric for greater safety
  • Soft vinyl cushions are removable for easy cleaning
  • Contoured one-piece headband follows shape of the head to minimize head pressure
  • Adjustable crown strap provides secure fit and allows choice of three positions: over-the-head, under-the-chin and behind-the-neck

Specifications

Color Red
ANSI S3.1974 Yes
Cushion Material Vinyl
Cushion Color Black
Headband Material Plastic
For Use With (Application) Agriculture, Automotive, Construction, Industry, Maintenance, Transportation, Utilities
High Visibility No
Model QM24PLUS
Noise Reduction Rating (NRR) 25 dB
Product Type Quiet Ear Muff
Headband Color Black
Dielectric Yes
Material ABS
Show More Show Less