Learn More
New England Biolab Ltd Hot Start Taq 2X Master Mix 500 reactions (50 l vol)
Catalog No. 50591070
Encompass
Hot Start Taq 2X Master Mix is an optimized ready-to-use solution containing Hot Start Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers. It is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It is recommended for amplification up to 5 kb. Hot Start Taq DNA Polymerase is a mixture of Taq DNA Polymerase and an aptamer-based inhibitor. The inhibitor binds reversibly to the enzyme, inhibiting polymerase activity at temperatures below 45C, but releases the enzyme during normal cycling conditions allowing reactions to be set up at room temperature. The aptamer-based hot start does not require a separate high temperature incubation step to activate the enzyme. Hot Start Taq DNA Polymerase possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity.
Catalog No. 50591070 Supplier New England Biolab Ltd Supplier No. M0496L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Hot Start Taq 2X Master Mix is an optimized ready-to-use solution containing Hot Start Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers. It is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It is recommended for amplification up to 5 kb. Hot Start Taq DNA Polymerase is a mixture of Taq DNA Polymerase and an aptamer-based inhibitor. The inhibitor binds reversibly to the enzyme, inhibiting polymerase activity at temperatures below 45C, but releases the enzyme during normal cycling conditions allowing reactions to be set up at room temperature. The aptamer-based hot start does not require a separate high temperature incubation step to activate the enzyme. Hot Start Taq DNA Polymerase possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity.