Learn More
Brady™ Hot Weather Self-Expiring Parking Permit
Monitor temporary parking with self-expiring parking permits. The Brady™ Hot Weather Temporary Self-Expiring Parking Permit is perfect for weather over 80°F/25°C.
Supplier: Brady™ 95677
Description
- Parking permits progressively expire throughout the day
- Reveals the letters E-X-P-I-R-E-D
- Expiration process starts once the self-adhesive cover is applied
- 500 per package
Specifications
| 5 in. | |
| 3 in. x 5 in. | |
| 3 in. |
| Red on white | |
| Permit parking |