missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences Hot Start Taq DNA Polymerase 2X Master Mix
Description
- G-biosciences Hot start Taq DNA Polymerase 2x master mix is ready to use premix which contains hot start Taq DNA Polymerase, dNTPs and optimized buffer with stabilizers.
- Hot start Taq DNA Polymerase is chemically modified that possesses a 5' - 3' polymerase activity and a 5'-flap endonuclease activity.
- Hot start Taq DNA Polymerase leads to complete inactivation of the polymerase until the initial heat activation step at the start of PCR.
- Hot start Taq DNA Polymerase reduces non-specific amplification during PCR setup and increases PCR specificity and sensitivity.
Specifications
Specifications
| Detection Method | Gel Electrophoresis |
| Form | Liquid |
| GC-Rich PCR Performance | High |
| PCR Method | Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR |
| Reaction Speed | Fast |
| Label or Dye | Biotin, 6-FAM, ROX, SYBR Green, TAMRA |
| Product Type | DNA Polymerase Master Mix |
| For Use With (Application) | Hot-start PCR, Multiplex PCR, High-fidelity PCR, Site-directed Mutagenesis, Next-Generation Sequencing (NGS) |
| No. of Reactions | 100 |