Learn More
Poltex Horizontal Tip Organizer-5 Pocket Magnet Mount
Poltex Horizontal Tip Organizer-5 Pocket is a horizontally mounted Pipette Tip Organizer for inside BSC, or other convenient area.
Supplier: Poltex HZORG5MBRK
Description
Horizontally mounted Pipette Tip Organizer for inside BSC, or other areas
Super durable and cleanable PETG construction
5 pockets
Optional add-ons:
- Pipette-Aide bracket
- 4 Pipettor bracket
- Both brackets
Magnets included to mount organizer
Specifications
| Pipette Aid Bracket | |
| 5 x 1 | |
| Clear | |
| PETG | |
| Rectangular | |
| Non-sterile |
| Pipette Tip Box Holder | |
| Not Autoclavable | |
| Pipette Tips | |
| 5 | |
| No |