missing translation for 'onlineSavingsMsg'
Learn More

Poltex Horizontal Tip Organizer

Catalog No. 22022567
Encompass
Change view
Click to view available options
Array:
1 x 3
1 x 4
1 x 5
Includes:
Pipette aid bracket
Pipettor and pipette aid brackets
Pipettor bracket
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array Includes
22022567 1 x 3 -
22022566 1 x 3 Pipette aid bracket
22022565 1 x 3 Pipettor and pipette aid brackets
22022564 1 x 3 Pipettor bracket
22022563 1 x 4 -
22022562 1 x 4 Pipette aid bracket
22022561 1 x 4 Pipettor and pipette aid brackets
22022560 1 x 4 Pipettor bracket
22022559 1 x 5 -
22022558 1 x 5 Pipette aid bracket
22022557 1 x 5 Pipettor and pipette aid brackets
22022556 1 x 5 Pipettor bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 22022567 Supplier Poltex Supplier No. HZORG3T
Only null left

Horizontally mounted Pipette Tip Organizer for use inside a BSC or other convenient area. Tape mount, comes with powerful VHB tape for mounting to stainless steel BSC surface.

  • Horizontally mounted pipette tip organizer for inside a BSC, or other convenient area
  • Includes powerful VHB tape for mounting to stainless steel BSC surface
  • Super durable and cleanable PETG construction.
  • Available in 3, 4, or 5 pocket
  • Choose from plain product, or with pipette-aide bracket, pipettor bracket, or both brackets

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipette Tips
Material PETG
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipette Tip Organizer
Length (Metric) 109.22 mm
Product Type Pipette Tip Box Holder
Array 1 x 3
Show More Show Less