Learn More
Poltex Horizontal Tip Organizer-4 Pocket with Magnet Mount
Catalog No. p-200089281
Change view
Click to view available options
Includes:
Both Brackets
None
Pipette Aid Bracket
Pipettor Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21100521 None
21100522 Pipette Aid Bracket
21100523 Both Brackets
21100524 Pipettor Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21100521 Supplier Poltex Supplier No. HZORG4M
Only null left
Poltex Horizontal Tip Organizer-4 Pocket is a horizontally mounted Pipette Tip Organizer for inside BSC, or other convenient area.
Horizontally mounted Pipette Tip Organizer for inside BSC, or other areas
Super durable and cleanable PETG construction
4 pockets
Optional add-ons:
  • Pipette-Aide bracket
  • 4 Pipettor bracket
  • Both brackets
Magnets included to mount organizer
Specifications
Autoclavable Not Autoclavable
Color Clear
Holds Pipette Tips
Material PETG
Positions 4
Shape Rectangular
Stackable No
Sterility Non-sterile
Includes None
Product Type Pipette Tip Box Holder
Array 4 x 1
Show More Show Less