missing translation for 'onlineSavingsMsg'
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Accessories: Reagent Cups

Catalog No. 23600506
Encompass
Change view
Click to view available options
Capacity (Metric):
10 mL
15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23600506 15 mL
23600504 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23600506 Supplier HORIBA Supplier No. 1221037307

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Pentra 400 Clinical Chemistry Analyzer

  • Available in 4, 10 and 15mL sizes

Specifications

Type Reagent Cup
Capacity (Metric) 15 mL