Learn More
Stoelting™ Hopkins Bulldog Clamp
Catalog No. 10000481
Change view
Click to view available options
Model:
Slight curve
Straight
Length (English):
1.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Length (English)
10000481 Straight 1.5 in.
10000482 Slight curve 1.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10000481 Supplier Stoelting™ Supplier No. 5212040P
Stoelting™ Hopkins Bulldog Clamp is useful in surgeries that require precise blood stoppage
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Specifications
For Use With (Application) Neuroscience
Length (English) 1.5 in.
Model Straight