Learn More
Stoelting™ Hopkins Bulldog Clamp
Stoelting™ Hopkins Bulldog Clamp is useful in surgeries that require precise blood stoppage
Supplier:  Stoelting™ 5212042P
 View more versions of this product
Catalog No. 10000482
Only null left
Description
Description
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Specifications
Chemical Identifiers
Neuroscience
Specifications
Neuroscience
1.5 in.
Slight curve
Product Suggestions
Product Suggestions