Learn More
Supertek™ Hooked Weight Set DFS Item
Hooked weight set consists of seven polished brass weights and two black enameled weights with flat countersunk bottoms. Each weight is accurately adjusted and the value is clearly indicated.
Supplier:  Supertek™ PH10190
Catalog No. S012342
Only null left
Description
Description
  • Hooked weight set of 9.
  • 7 polished brass weights and 2 black enameled weights.
  • Each weight is accurately adjusted and the value is clearly indicated.
  • Individual masses are as follows: 1 x 10 g, 2 x 20 g, 1 x 50 g, 1 x 100 g, 2 x 200 g, 1 x 500 g, 1 x 1000 g
Specifications
Specifications
Mass Set
Brass, Iron
10 g, 20 g, 50 g, 100 g, 200 g, 500 g, 1000 g
Set of 7
Product Suggestions
Product Suggestions