missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Hooked Cylinder

Catalog No. S12921
Change view
Click to view available options
Material:
Aluminum
Brass
Copper
Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S12921 Copper
S12918 Aluminum
S12919 Steel
S12920 Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S12921 Supplier Eisco™ Supplier No. PH0112E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hooked Cylinder for use with physics experiments.

  • Can be used for specific heat, specific gravity or density.
  • Used to study concepts related to physics.

Specifications

Grade 7 to 12
Dimensions (L x W) 5 cm x 13 mm
For Use With (Application) For use with physics experiments
For Use With (Equipment) Used for specific heat, specific gravity, density
Includes 1 Cylinder
Material Copper
Class Size 1 Lab Group
Product Type Hooked Cylinder