Learn More
Eisco™ Hooke's Law Apparatus for Student Use - Supplied With Weights
Eisco Labs Hooke's Law Apparatus for student use. Supplied with weights, ideal for demonstrating the principles of elasticity and Hooke's Law in educational settings. Perfect for hands-on learning and experiments.
Supplier:  Eisco™ PH0323A
Catalog No. S02910
Only null left
Explore available promotions
Description
Description
  • Complete apparatus has a 15 cm adjustable scale, marked in mm, to help prevent parallax error
  • Scale is mounted on sturdy 30 cm support rod
  • Apparatus is situated on a sturdy wooden base
  • Includes 9 slotted masses and hanger, each weighing 10g, for a total of 100g
  • Ideal for demonstrating Hooke's Law which states the force needed to extend or compress a spring by some distance is proportional to that distance
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
Hooke's Law Apparatus
Product Suggestions
Product Suggestions