missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Hood Flashlight with Mounting Kit

Catalog No. 17001748
Encompass
Change view
Click to view available options
Flourescent Yellow and Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17001748 Flourescent Yellow and Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17001748 Supplier National Safety Apparel™ Supplier No. HLITEKIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mounts to lift front hoods, increasing visibility in hazard areas

  • For use with lift front hoods
  • Intrinsically safe light, Class 1 Div 1
  • 60 Lumens
  • Uses 2 AAA batteries (included)
  • Push-button switch for easy one handed operation
  • Light stays in place when shield is lifted
  • Clips on right or left side Imported

Specifications

Size Medium
Material Plastic
Color Black