missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Thermostat™ Cold Weather Glove Liners

Catalog No. 191201206
Change view
Click to view available options
:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
191201206 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 191201206 Supplier Honeywell™ Supplier No. TH13A BL
Only null left

13 gauge Thermostat™ fiber has a hollow core which traps air for greater thermal protection. Seamless knit construction prevents rubbing or chafing to provide workers with enhanced comfort and warmth.

  • 13 gauge seamless knit, Thermostat™ glove liners
  • Thermostat™ fiber has a hollow core to trap air for greater thermal protection
  • Seamless knit construction prevents rubbing or chafing
  • Ideal liner for cold weather applications

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color Blue
Hand Ambidextrous
Material Thermostat Fiber
Size Universal
For Use With (Application) Refrigeration, Assembly/Parts Handling, General Maintenance, and Frozen Food Warehousing
Glove Cuff Style Knit
Series TH13A
Yarn Gauge 13