missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Fendall Saline Refill Bottles for Wall Stations

Catalog No. 19003030
Change view
Click to view available options
Capacity (Metric):
473.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19003030 473.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19003030 Supplier Honeywell™ Supplier No. 320004540000H5
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use as refills for wall-mounted stations or individually

  • Easy-open top
  • User-controlled flush volume
  • 36-month shelf life
  • Works with most wall-mount eyewash stations

Specifications

Capacity (English) 16 oz.
Capacity (Metric) 473.1 mL
For Use With (Equipment) Fendall Wall Station
Product Type Refill Solution