Learn More
Honeywell™ Sperian™ Fendall™ Refill Cartridges for Emergency Eyewash Stations
Catalog No. 172922B
Change view
Click to view available options
For Use With (Equipment):
EyeSaline Flashflood Eyewash Station
Catalog No. For Use With (Equipment)
172922B EyeSaline Flashflood Eyewash Station
1 options
Catalog No. 172922B Supplier Honeywell™ Supplier No. 320004010000
Offers unmatched safety in the workplace. Sperian™ Honeywell™ Fendall™ Refill Cartridges for Emergency Eyewash Stations deliver a preserved, buffered, pH balanced solution.
Offers unmatched safety in the workplace. Sperian™ Honeywell™ Fendall™ Refill Cartridges for Emergency Eyewash Stations deliver a preserved, buffered, pH balanced solution.
  • Easy to install
  • Delivers three minutes of solution
  • Up to 24-month shelf life
  • Cartridges hold 3.8L of solution
Order Info
In packs of four cartridges.
Specifications
Capacity (English) 128 oz.
Capacity (Metric) 3785 mL
For Use With (Equipment) EyeSaline Flashflood Eyewash Station
Product Type Refill Cartridge