Learn More
Honeywell™ PowerCoat™ Disposable Tri-Polymer Gloves
Catalog No. 191301847C
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191301847C Large
191301847A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 191301847C Supplier Honeywell™ Supplier No. PSDTRIPL
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Powder free gloves are constructed from a specialty blend of latex, neoprene, and nitrile to offer protection from chemicals and liquids. A rough finish on the fingertips enhances grip in wet applications.
  • Tri-polymer disposable gloves are constructed from a combination of latex, neoprene and nitrile
  • Rough finish on the fingertips enhances grip in wet applications
  • Powder free design for reduced product contamination
  • Ideal for use in Agriculture, Chemical, and Oil/Gas Industries
Specifications
Color Purple
Disposable Single-use
Exterior Finish Partially Textured
Hand Ambidextrous
Latex Free No
Material Latex, Nitrile, Neoprene
Powdered Powder Free
Size Large
Thickness (English) 5 mil
Thickness (Metric) 0.13 mm
Series PSD-TRIP
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less