missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ NorthFlex Light Task™ ESD Anti-Static Conductive Gloves

Catalog No. 19166714
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19166714 Small
19166715 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19166714 Supplier Honeywell™ North™ Supplier No. NF15ESD/7S

Lightweight, 15-gauge liner is constructed of copper based Thunderon™, with low surface resistivity when tested in accordance with EN 1149.

  • 15-gauge liner constructed of nylon and copper-based Thunderon™
  • Polyurethane coating on the palm and fingertips for enhanced grip capabilites
  • Low surface resistivity (<4 x 10/sq.) when tested in accordance with EN 1149

Specifications

Certifications/Compliance ANSI/ISEA 105, ASTM D3389-05
Certifications/Compliance (Europe) EN 1149
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish Partially Textured
Hand Specific
Material Nylon, Thunderon
Product Type ESD Gloves
Size Small
For Use With (Application) Electronic Component Assembly/Handling
Glove Cuff Style Knit
Series NF15ESD
Yarn Gauge 15
Length (English) 20.5 in.
Length (Metric) 52 cm
Show More Show Less