Learn More
Honeywell™ North™ Silver Shield™ Chemical Resistant Aprons
Description
- Five layer PE and EVOH constructions provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
- Provides high performance protection in the most demanding applications
Specifications
Specifications
| Type | Chemical-resistant Apron |
| Size | Universal |
| Material | Composite Fabric |
| Color | Silver |
| Includes | No Sleeves |