missing translation for 'onlineSavingsMsg'
Learn More

Homopiperazine 98.0+%, TCI America™

Catalog No. h057225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
H057225G 25 g
1 options

Catalog No. H057225G

Supplier: TCI America H057225G

Only null left

Chemical Identifiers

CAS 505-66-8
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00006933
InChI Key FQUYSHZXSKYCSY-UHFFFAOYSA-N
Synonym homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine
PubChem CID 68163
IUPAC Name 1,4-diazepane
SMILES C1CNCCNC1

Specifications

Melting Point 45°C
Color White
Boiling Point 167°C
Quantity 25 g
UN Number 1759
Formula Weight 100.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material Homopiperazine
RTECSNumber HM3850000
TSCA Yes