Learn More
Homopiperazine, 98%
CAS: 505-66-8 | C5H12N2 | 100.165 g/mol
$220.66 - $612.56
Chemical Identifiers
| CAS | 505-66-8 |
|---|---|
| Molecular Formula | C5H12N2 |
| Molecular Weight (g/mol) | 100.165 |
| MDL Number | MFCD00006933 |
| InChI Key | FQUYSHZXSKYCSY-UHFFFAOYSA-N |
| Synonym | homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine |
| PubChem CID | 68163 |
| IUPAC Name | 1,4-diazepane |
| SMILES | C1CNCCNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1575114
|
Thermo Scientific Chemicals
A1575114 |
25 g |
Each for $220.66
|
|
|||||
|
AAA1575122
|
Thermo Scientific Chemicals
A1575122 |
100 g |
Each for $612.56
|
|
|||||
Description
Homopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHomopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.
Solubility
Soluble in water.
Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
| 505-66-8 | |
| 100.165 | |
| FQUYSHZXSKYCSY-UHFFFAOYSA-N | |
| 68163 | |
| C1CNCCNC1 |
| C5H12N2 | |
| MFCD00006933 | |
| homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine | |
| 1,4-diazepane |
Specifications
| 505-66-8 | |
| 0.95 | |
| 64°C (147°F) | |
| C5H12N2 | |
| 25 g | |
| 102711 | |
| homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine | |
| FQUYSHZXSKYCSY-UHFFFAOYSA-N | |
| 1,4-diazepane | |
| 68163 | |
| 98% |
| 38°C to 44°C | |
| 168°C to 170°C | |
| Amine-like | |
| MFCD00006933 | |
| UN3259 | |
| Air sensitive; Hygroscopic | |
| Soluble in water. | |
| C1CNCCNC1 | |
| 100.165 | |
| 100.17 | |
| Homopiperazine |
Safety and Handling
GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H312-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber : 208-016-1
RTECSNumber : HM3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only