missing translation for 'onlineSavingsMsg'
Learn More

Homopiperazine, 98%

Catalog No. AAA1575122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1575122 100 g
AAA1575114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1575122 Supplier Thermo Scientific Chemicals Supplier No. A1575122

CAS: 505-66-8 | C5H12N2 | 100.165 g/mol

Homopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Homopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 505-66-8
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00006933
InChI Key FQUYSHZXSKYCSY-UHFFFAOYSA-N
Synonym homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine
PubChem CID 68163
IUPAC Name 1,4-diazepane
SMILES C1CNCCNC1

Specifications

Melting Point 38°C to 44°C
Density 0.95
Boiling Point 168°C to 170°C
Flash Point 64°C (147°F)
Odor Amine-like
Quantity 100 g
UN Number UN3259
Beilstein 102711
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
Formula Weight 100.17
Percent Purity 98%
Chemical Name or Material Homopiperazine
Show More Show Less
Hazard Category H312-H314
Hazard Statement GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 208-016-1
RTECSNumber HM3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only