missing translation for 'onlineSavingsMsg'
Learn More

Homopiperazin-5-one, 95%

Catalog No. AAH26990MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26990MD 250 mg
AAH2699003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26990MD Supplier Thermo Scientific Chemicals Supplier No. H26990MD
Only null left

CAS: 34376-54-0 | C5H10N2O | 114.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34376-54-0
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.148
MDL Number MFCD00766804
InChI Key QPPLBCQXWDBQFS-UHFFFAOYSA-N
Synonym 1,4 diazepan-5-one, homopiperazin-5-one, 5-homopiperazinone, 2,3,6,7-tetrahydro-1h-1,4-diazepin-5 4h-one, 5h-1,4-diazepin-5-one, hexahydro, 2,3,4,5,6,7-hexahydro-1h-1,4-diazepin-5-one, peakdale1_000765, acmc-1ctxs, 1,4-diazepan-5-one, hexahydro-1h-1,4-diazepin-5-one
PubChem CID 2737264
IUPAC Name 1,4-diazepan-5-one
SMILES C1CNCCNC1=O

Specifications

Quantity 250 mg
Formula Weight 114.14
Percent Purity 95%
Chemical Name or Material Homopiperazin-5-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only