missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homophthalic Anhydride 98.0+%, TCI America™
$125.13 - $417.77
Chemical Identifiers
| CAS | 703-59-3 |
|---|---|
| Molecular Formula | C9H6O3 |
| Molecular Weight (g/mol) | 162.144 |
| MDL Number | MFCD00006894 |
| InChI Key | AKHSBAVQPIRVAG-UHFFFAOYSA-N |
| Synonym | homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride |
| PubChem CID | 12801 |
| IUPAC Name | 4H-isochromene-1,3-dione |
| SMILES | C1C2=CC=CC=C2C(=O)OC1=O |
Chemical Identifiers
| 703-59-3 | |
| 162.144 | |
| AKHSBAVQPIRVAG-UHFFFAOYSA-N | |
| 12801 | |
| C1C2=CC=CC=C2C(=O)OC1=O |
| C9H6O3 | |
| MFCD00006894 | |
| homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride | |
| 4H-isochromene-1,3-dione |
Specifications
| 703-59-3 | |
| White-Yellow | |
| C9H6O3 | |
| 5 g | |
| AKHSBAVQPIRVAG-UHFFFAOYSA-N | |
| 4H-isochromene-1,3-dione | |
| 12801 | |
| ≥98.0% (GC) | |
| Homophthalic Anhydride |
| 142°C | |
| 161°C | |
| MFCD00006894 | |
| homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride | |
| C1C2=CC=CC=C2C(=O)OC1=O | |
| 162.144 | |
| 162.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No