missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic anhydride, 97+%

Catalog No. AAA1553809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1553809 10 g
AAA1553818 50 g
AAA1553830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1553809 Supplier Thermo Scientific Chemicals Supplier No. A1553809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 703-59-3 | C9H6O3 | 162.144 g/mol

It is used as a chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chemical and organic intermediates.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Storage temperature 15 - 30°C. Store under inert gas. Store away from exposure to moisture, strong oxidizing agents, strong acids, strong bases.

Chemical Identifiers

CAS 703-59-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.144
MDL Number MFCD00006894
InChI Key AKHSBAVQPIRVAG-UHFFFAOYSA-N
Synonym homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride
PubChem CID 12801
IUPAC Name 4H-isochromene-1,3-dione
SMILES C1C2=CC=CC=C2C(=O)OC1=O

Specifications

Melting Point 140°C to 144°C
Quantity 10 g
Beilstein 5244
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 162.14
Percent Purity ≥97%
Chemical Name or Material Homophthalic anhydride
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-873-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only