missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 89-51-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00004326
InChI Key ZHQLTKAVLJKSKR-UHFFFAOYSA-N
Synonym homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
PubChem CID 66643
IUPAC Name 2-(carboxymethyl)benzoic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120840250
SDS View Documents
Thermo Scientific Chemicals
120840250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC120841000
SDS View Documents
Thermo Scientific Chemicals
120841000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

89-51-0
180.16
ZHQLTKAVLJKSKR-UHFFFAOYSA-N
66643
C1=CC=C(C(=C1)CC(=O)O)C(=O)O
C9H8O4
MFCD00004326
homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
2-(carboxymethyl)benzoic acid

Specifications

89-51-0
97.5% min. (HPLC)
C9H8O4
MFCD00004326
09, 857
Solubility in water: 12g/L (20 c)
C1=CC=C(C(=C1)CC(=O)O)C(=O)O
180.16
180.16
Homophthalic acid, 99%
Authentic
Glass bottle
HO2CCH2C6H4CO2H
25g
homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
ZHQLTKAVLJKSKR-UHFFFAOYSA-N
2-(carboxymethyl)benzoic acid
66643
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 201-913-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only