missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homomorpholine hydrochloride, 98%
Description
Homomorpholine Hydrochloride is a useful synthetic intermediate. It is used to prepare homomorpholine oxazolidinone with antibacterial activities. It is also used to synthesize A2A adenosine receptor antagonists for the treatment of Parkinson's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Slightly soluble in water.
Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 158°C to 164°C |
| Quantity | 1 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 137.61 |
| Percent Purity | 98% |
| Chemical Name or Material | Homomorpholine hydrochloride |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
RUO – Research Use Only