missing translation for 'onlineSavingsMsg'
Learn More

Homomorpholine hydrochloride, 98%

Catalog No. AAB2174906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2174906 5 g
AAB2174903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2174906 Supplier Thermo Scientific Chemicals Supplier No. B2174906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 178312-62-4 | C5H12ClNO | 137.607 g/mol

Homomorpholine Hydrochloride is a useful synthetic intermediate. It is used to prepare homomorpholine oxazolidinone with antibacterial activities. It is also used to synthesize A2A adenosine receptor antagonists for the treatment of Parkinson's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 178312-62-4
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD02683070
InChI Key LJRCWNIWOVZLKS-UHFFFAOYSA-N
Synonym 1,4-oxazepane hydrochloride, homomorpholine hydrochloride, 1,4-oxazepine, hexahydro-, hydrochloride, homomorpholine hcl, 1,4-oxazepan hydrochloride, 1,4 oxazepane hydrochloride, hexahydro-1,4-oxazepine hydrochloride, homomorpholine-hydrochloride, homo-morpholine hydrochloride, homomorph-oline hydrochloride
PubChem CID 23090214
IUPAC Name 1,4-oxazepane;hydrochloride
SMILES C1CNCCOC1.Cl

Specifications

Melting Point 158°C to 164°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 137.61
Percent Purity 98%
Chemical Name or Material Homomorpholine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only