missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homatropine Hydrochloride 98.0+%, TCI America™
$68.97 - $536.84
Chemical Identifiers
| CAS | 637-21-8 |
|---|---|
| Molecular Formula | C16H22ClNO3 |
| Molecular Weight (g/mol) | 311.806 |
| MDL Number | MFCD00050613 |
| InChI Key | FRMDDIJBLQNNTC-ISXUQMITSA-N |
| PubChem CID | 76966341 |
| IUPAC Name | [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;hydrochloride |
| SMILES | CN1C2CCC1CC(C2)OC(=O)C(C3=CC=CC=C3)O.Cl |
Chemical Identifiers
| 637-21-8 | |
| 311.806 | |
| FRMDDIJBLQNNTC-ISXUQMITSA-N | |
| [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;hydrochloride |
| C16H22ClNO3 | |
| MFCD00050613 | |
| 76966341 | |
| CN1C2CCC1CC(C2)OC(=O)C(C3=CC=CC=C3)O.Cl |
Specifications
| 637-21-8 | |
| White | |
| MFCD00050613 | |
| 1544 | |
| CN1C2CCC1CC(C2)OC(=O)C(C3=CC=CC=C3)O.Cl | |
| 311.806 | |
| 311.81 | |
| Crystalline Powder |
| 220°C | |
| C16H22ClNO3 | |
| 1 g | |
| FRMDDIJBLQNNTC-ISXUQMITSA-N | |
| [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;hydrochloride | |
| 76966341 | |
| ≥98.0% (T) | |
| Homatropine Hydrochloride |
Safety and Handling
TSCA : No