missing translation for 'onlineSavingsMsg'
Learn More

Homatropine Hydrobromide 99.0+%, TCI America™

Catalog No. H01551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H01551G Supplier TCI America Supplier No. H01551G

Specifications

CAS 51-56-9
Melting Point 212°C
Color White
Molecular Formula C16H22BrNO3
Quantity 1 g
MDL Number MFCD00012676
UN Number 1544
Synonym homatropine bromide, unii-bew7469qz0 component dwsgtftvblxelc-uoizbmalsa-n
InChI Key DWSGTFTVBLXELC-MYKIJENHSA-N
SMILES CN1C2CCC1CC(C2)OC(=O)C(C3=CC=CC=C3)O.Br
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2S)-2-hydroxy-2-phenylacetate;hydrobromide
Molecular Weight (g/mol) 356.26
PubChem CID 73415930
Formula Weight 356.26
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Homatropine Hydrobromide
Show More Show Less
RTECSNumber YM5602000
TSCA Yes