missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Catalog No. AA40575NC
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40575NC 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40575NC Supplier Thermo Scientific Chemicals Supplier No. 040575NC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14017-54-0 | Cl3HoO12 | 463.268 g/mol

Holmium(III) perchlorate is used to verify the wavelength scale of UV/visible spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14017-54-0
Molecular Formula Cl3HoO12
Molecular Weight (g/mol) 463.268
MDL Number MFCD00064643
InChI Key HRWMCDDQLSZQPE-UHFFFAOYSA-K
Synonym holmium perchlorate, acmc-20akr8, holmium iii perchlorate solution, perchloric acid holmium iii salt, holmium 3+ triperchlorate, perchloric acid,holmium 3+ salt 8ci,9ci
PubChem CID 14345154
IUPAC Name holmium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ho+3]

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Soluble in water.
Formula Weight 571.37 (463.28 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Density 1.57 g/mL
Chemical Name or Material Holmium(III) perchlorate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-839-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only