missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)

CAS: 14483-18-2 | H10HoN3O14 | 441.017 g/mol

$203.01 - $203.01

Chemical Identifiers

CAS 14483-18-2
Molecular Formula H10HoN3O14
Molecular Weight (g/mol) 441.017
MDL Number MFCD00149737
InChI Key BJUGWWDCFYEYOA-UHFFFAOYSA-N
Synonym holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
PubChem CID 25021754
IUPAC Name holmium(3+);trinitrate;pentahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1458809
View Documents
Thermo Scientific Chemicals
01458809
10 g
Each for $203.01
Only null left
 
Description

Description

Holmium(III) nitrate pentahydrate is used to prepare holmium-beta-cyclodextrin complex by reacting with beta- cyclodextrin, which posses potential pharmacological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14483-18-2
441.017
BJUGWWDCFYEYOA-UHFFFAOYSA-N
25021754
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]
H10HoN3O14
MFCD00149737
holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
holmium(3+);trinitrate;pentahydrate

Specifications

14483-18-2
10 g
MFCD00149737
holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
BJUGWWDCFYEYOA-UHFFFAOYSA-N
holmium(3+);trinitrate;pentahydrate
25021754
99.9%
Odorless
Holmium(III) nitrate pentahydrate
Crystalline Aggregates
H10HoN3O14
UN1477
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]
441.017
441.02 (350.94 Anhydrous)
Reacton
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

EINECSNumber : 233-438-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only