missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Catalog No. p-7049202
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1319904 2 g
AA1319909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1319904 Supplier Thermo Scientific Chemicals Supplier No. 01319904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14589-33-4 | C15H21HoO6 | 462.26 g/mol

It is used for Building Blocks of chemicals and , Chemical Synthesis, used as a trihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14589-33-4
Molecular Formula C15H21HoO6
Molecular Weight (g/mol) 462.26
MDL Number MFCD00050176
InChI Key UEKRGRZSLATUQV-UHFFFAOYSA-N
Synonym holmium 2,4-pentanedionate, holmium iii 2,4-pentanedionate, reacton, holmium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy holmio oxy pent-3-en-2-one
PubChem CID 24282829
IUPAC Name holmium;(Z)-4-hydroxypent-3-en-2-one
SMILES [Ho+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 2 g
Odor Odorless
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 462.26
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Holmium(III) 2,4-pentanedionate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only