Learn More
Corning™ PYREX™ Hollow Glass Standard Taper Joint Stopper
GREENER_CHOICE
Catalog No. p-78279
Change view
Click to view available options
Standard Taper Joint:
19/38
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
14640J 24/40
14640H 19/38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14640J Supplier Corning™ Supplier No. 7570N24
Only null left
Hollow stoppers, closed at bottom, conform to ASTM™ E 675-94
These hollow full length PYREX™ standard taper joint stoppers are closed at the bottom. They are interchangeable with standard taper joints of the same size. These hollow stoppers are light in weight, yet very strong to reduce chipping and breakage, thus minimizing replacement costs. A barrel-shaped head means it is easy to clean and even easier to rotate in or out. Grooves in the sides help prevent slippage. The flat top allows the stopper to stand on its head, which frees the user's hand and minimizes potential contamination.
Specifications
Standard Taper Joint 24/40
Product Type Barrelhead Stopper
Material Borosilicate Glass