Learn More
Eisco™ Hollow Equilateral Prism, 45 mm
Catalog No. S27718
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27718 1 Ea.
1 options
Catalog No. S27718 Supplier Eisco™ Supplier No. PH0575
Hollow Equilateral Acrylic Prism has one hole for adding fluids to study diffraction and dispersion
  • Acrylic Equilateral Prism with 45 mm sides
  • Has one hole for adding fluids to study diffraction and dispersion
  • Supplied with a funnel and stopper
Specifications
Product Type Prism, Hollow Equilateral
Description 45 mm Sides
Material Acrylic
For Use With (Application) Studying Diffraction and Dispersion
Includes Funnel, Stopper
Quantity 1 Ea.