Learn More
Electron Microscopy Sciences EMS Holey Carbon Film 50 m holes on 300 mesh Copper
Catalog No. 5024446
Encompass
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Holes: 50 m
Catalog No. 5024446 Supplier Electron Microscopy Sciences Supplier No. HC300CU
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Holes: 50 m