missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Petroleum Distillate, Spectrum™ Chemical

Catalog No. 18611159
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611159 500 mL Amber Glass Bottle
18611155 1 L Amber Glass Bottle
18611156 4 L Amber Glass Bottle
18611157 20 L Amber Glass Bottle
18611158 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611159 Supplier Spectrum Chemical Mfg Cor Supplier No. K1025500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8008-20-6

Specifications

CAS Max % 1
Boiling Range 151° to 301°C
Boiling Point 151°C to 301°C/303.8°F to 573.8°F
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Kerosene, Petroleum Distillate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.