Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Catalog No. 361013415
Change view
Click to view available options
Product Type:
Disposable Blade Carrier EC
Disposable Blade Carrier EC70
Disposable Blade Carrier EV
Standard Knife Holder
For Use With (Equipment):
HM 525 NX and HM 550
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
361013415 Standard Knife Holder HM 525 NX and HM 550
361013416 Disposable Blade Carrier EC HM 525 NX and HM 550
361013419 Disposable Blade Carrier EV HM 525 NX and HM 550
361016411 Disposable Blade Carrier EC70 HM 525 NX and HM 550
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361013415 Supplier Epredia™ Supplier No. 705460
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
Standard Knife Carrier:
  • Allows for insertion of standard knives, with clearance angle adjustment
  • Can be moved horizontally on precision dovetail guide with quick clamp system
  • Includes anti-roll plate
Disposable Blade Carrier EC:
  • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
  • Can be moved on precision dovetail guide with quick clamp plate
  • Includes anti-roll plate
Disposable Blade Carrier EV:
  • Use with vacutome system
  • Allows for insertion of high- and low-profile disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
  • Use with vacutome system
  • Allows for insertion of disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Specifications
Product Type Standard Knife Holder
For Use With (Equipment) HM 525 NX and HM 550