missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One HLA Terasaki 72-well Plates

Catalog No. 07001026
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07001026 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07001026 Supplier Greiner Bio-One Supplier No. 769190P

Tissue-cultured treated plates.

  • Made from polystyrene
  • Recessed lid design
  • Clear

Specifications

Description HLA Terasaki plate
Color Clear
Product Type Multi-well Plate
Material Polystyrene
No. of Wells 72
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)