missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Phenyl (High Sub)

Catalog No. 12101333
Change view
Click to view available options
Bed Volume:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
12101333 5 mL
12101334 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12101333 Supplier Cytiva Supplier No. 17545109

Hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.

Capto Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification

  • Aromatic ligand coupled with a flexible spacer for laboratory and process applications requiring a HIC resin with resin to high hydrophobicity
  • Highly rigid matrix enables high flow rates, long bed heights and faster throughput
  • Improves productivity and process economy in downstream operations
  • Narrow specification range ensures robust performance in sensitive processes
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide

Specifications

Bed Volume 5 mL
Diameter (Metric) Inner 16 mm
Product Type Hydrophobic Interaction Chromatography (HIC) Selection Kit
Flow Rate 600 cm/hr.
Quantity 5/Pk.
Ligand Type Phenyl
Material Agarose
Particle Size 75 μm
pH Range 3 to 13
Content And Storage 4°C to 30°C
Binding Capacity 27 mg/mL
Height (Metric) Bed 25 mm
Show More Show Less