missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Octyl HIC Column

Catalog No. 12101330
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12101330 1 mL
12101329 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12101330 Supplier Cytiva Supplier No. 17546508

Prepacked with Capto™ Octyl hydrophobic interaction chromatography (HIC) resin for small scale purification.

HiTrap™ Capto™ Octyl column is prepacked with a HIC resin for small scale purification

  • Prepacked with Capto™ Octyl high-flow and high-capacity HIC resin for capture and intermediate purification processes
  • Lab-scale column format, suitable for screening of selectivity, binding and elution conditions, and for small-scale purifications in research and process development
  • Convenient HiTrap™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA™ system
  • Manufactured from biocompatible polypropylene that does not interact with biomolecules
  • Has an octyl ligand for the capture and intermediate purification of monoclonal antibodies (mAbs), mAb variants, and other biomolecules

Specifications

Bed Volume 1 mL
Diameter (Metric) Inner 7 mm
Product Type HiTrap Octyl FF
Flow Rate 1 mL/min.
Quantity 5/Pk.
Ligand Type Octyl
Max. Pressure 3 bar
Particle Size 75 μm
pH Range 3 to 13
Content And Storage 4°C to 30°C
Storage Buffer 20% Ethanol
Binding Capacity 25 mg
Height (Metric) Bed 25 mm
Capacity (Metric) 1 mL
Show More Show Less